
1,2-Diiodobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-42-9
:
Formula:C₆H₄I₂
MDL Number:MFCD00001038
:329.91 g/mol
Boiling Pt:282 °C
Density:2.524
Flash Pt:>110 °C (230 °F)
EINECS:210-425-5
:GHS07
Storage Temperature:Ambient