Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Diiodobenzene
1,2-Diiodobenzene 98%
1,2-Diiodobenzene 98%
:
Thermo Fisher Scientific
1,2-Diiodobenzene 98%
:
Thermo Fisher Scientific
:
615-42-9
:
:
C₆H₄I₂
:
MFCD00001038
:
329.91 g/mol
:
152 °C (15 mmHg)
:
2.52 g/cm³
:
>110 °C
:
210-425-5
:
GHS07
:
GHS05
:
Refrigerator
Appearance:
Clear yellow to brown Liquid
Infrared spectrum:
Conforms
GC:
≥97.5 %
ACRO21840