

4-Fluorothiophenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:371-42-6
:
Formula:C₆H₅FS
MDL Number:MFCD00004846
:128.17 g/mol
Boiling Pt:164 °C (1013 hPa)
Density:1,1996 g/cm³ (20 °C)
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:206-737-6
UN:1228
ADR:3,III
:GHS06
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5490 to 1.5520 (20°C, 589 nm)
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure