2,6-Difluorophenol
Catalog # MOLE10189897-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:66 °C (20 mmHg)
Melting Pt:46 °C
Density:1.27 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:130.09 g/mol
EINECS:248-884-9
UN:2921
CAS Number:28177-48-2
Formula:C₆H₄F₂O
MDL Number:MFCD00002158