


2,6-Difluorophenol 98%
Catalog # ACRO311630050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless Low melting crystalline solid
- Infrared spectrum:Conforms
- Melting point:38 to 42 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
Boiling Pt:59…61 °C (17 mmHg)
Melting Pt:38…42 °C
Density:1,27 g/cm³ (20 °C)
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:248-884-9
UN:2925
ADR:4.1,III
:130.09 g/mol
:GHS07
:GHS05
:GHS02
CAS Number:28177-48-2
Formula:C₆H₄F₂O
MDL Number:MFCD00002158