Order Entry
Export
ContactUsLinkComponent
3,4-Difluorophenol
Irritant
3,4-Difluorophenol
Catalog # MOLEM68493986
Supplier:  Molekula
CAS Number:  
Irritant
3,4-Difluorophenol
Catalog # MOLEM68493986
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :130.09 g/mol
Boiling Pt:185 °C (1013 hPa)
Melting Pt:32…36 °C
UN:2921
ADR:8,II
 :GHS07
CAS Number:2713-33-9
Formula:C₆H₄F₂O
MDL Number:MFCD00010315