Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluorophenol
3,4-Difluorophenol
3,4-Difluorophenol
Irritant
3,4-Difluorophenol
Supplier:
Thermo Scientific
Certificates
Expand image
Irritant
3,4-Difluorophenol
Supplier:
Thermo Scientific
About this item
CAS Number:
2713-33-9
:
Formula:
C₆H₄F₂O
MDL Number:
MFCD00010315
:
130.09 g/mol
Boiling Pt:
185 °C (1013 hPa)
Melting Pt:
32…36 °C
UN:
2921
ADR:
8,II
:
GHS07