2,3-Difluorophenol
Glass bottle for solids, 5 g
Catalog # MOLE48747018-5G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:6418-38-8
- Formula:C₆H₄F₂O
- MDL Number:MFCD00010262
- Molecular Weight:130.09 g/mol
- Boiling Pt:54 °C (25 mmHg)
- Melting Pt:30…32 °C
- UN:2921
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



