2,3-Difluorophenol ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 6418-38-8 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Odour: Phenol-like Boiling Point: 53-55°/25mm Beilstein Reference: 2082265 UN No.: 2921 MDL No.: MFCD00010262
- CAS Number:6418-38-8
- Formula:C₆H₄F₂O
- MDL Number:MFCD00010262
- Molecular Weight:130.09 g/mol
- Boiling Pt:53…55 °C (25 mmHg)
- Melting Pt:34…38 °C
- Flash Pt:56 °C (132 °F)
- Storage Temperature:Ambient
- UN:2921
- ADR:8,II
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 6418-38-8 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 56°(132°F) Odour: Phenol-like Boiling Point: 53-55°/25mm Beilstein Reference: 2082265 UN No.: 2921 MDL No.: MFCD00010262
- CAS Number:6418-38-8
- Formula:C₆H₄F₂O
- MDL Number:MFCD00010262
- Molecular Weight:130.09 g/mol
- Boiling Pt:53…55 °C (25 mmHg)
- Melting Pt:34…38 °C
- Flash Pt:56 °C (132 °F)
- Storage Temperature:Ambient
- UN:2921
- ADR:8,II



