1,4-Difluoro-2-nitrobenzene
Glass bottle for solids, 100 g
Catalog # MOLEM13564932
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,5-Difluoronitrobenzene
- CAS Number:364-74-9
- Formula:C₆H₃F₂NO₂
- MDL Number:MFCD00007054
- Molecular Weight:159.09 g/mol
- Boiling Pt:207 °C (1013 hPa)
- Melting Pt:–11.7 °C
- Density:1.4671 g/cm³ (17 °C)
- EINECS:206-663-4
- UN:2810
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



