Order Entry
Export
ContactUsLinkComponent
1,3-Difluoro-4-nitrobenzene
1,3-Difluoro-4-nitrobenzene
Catalog # MOLEM55383446
Supplier:  Molekula
CAS Number:  
1,3-Difluoro-4-nitrobenzene
Catalog # MOLEM55383446
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :159.09 g/mol
Boiling Pt:203 °C (1013 hPa)
Melting Pt:10 °C
Density:1.4571 g/cm³ (14 °C)
EINECS:207-167-0
UN:2810
ADR:6.1,III
CAS Number:446-35-5
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00007050