


1,2-Difluoro-4-nitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:369-34-6
:
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00007198
:159.09 g/mol
Boiling Pt:76…80 °C (8 mmHg)
Density:1.441
Flash Pt:80 °C (176 °F)
EINECS:206-718-2
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient