3-Fluoroaniline 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:372-19-0
- Formula:C₆H₆FN
- MDL Number:MFCD00007758
- Molecular Weight:111.12 g/mol
- Boiling Pt:186 °C (756 mmHg)
- Melting Pt:–2 °C
- Density:1.15 g/cm³
- Flash Pt:77 °C
- Storage Temperature:Ambient
- EINECS:206-747-0
- UN:2941
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:372-19-0
- Formula:C₆H₆FN
- MDL Number:MFCD00007758
- Molecular Weight:111.12 g/mol
- Boiling Pt:186 °C (756 mmHg)
- Melting Pt:–2 °C
- Density:1.15 g/cm³
- Flash Pt:77 °C
- Storage Temperature:Ambient
- EINECS:206-747-0
- UN:2941
- ADR:6.1,III
Documents
ACRO11928
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5430 to 1.5450 (20°C, 589 nm)
- GC:≥97.5 %



