3,5-Difluoroaniline
Glass bottle for solids, 5 g
Catalog # MOLEM69684741
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:372-39-4
- Formula:C₆H₅F₂N
- MDL Number:MFCD00007763
- Molecular Weight:129.11 g/mol
- Boiling Pt:185 °C (1013 hPa)
- Melting Pt:37…41 °C
- Density:1.295 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-752-8
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



