Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoroaniline
2,6-Difluoroaniline
2,6-Difluoroaniline
:
Thermo Scientific
2,6-Difluoroaniline
:
Thermo Scientific
:
5509-65-9
:
:
C₆H₅F₂N
:
MFCD00007655
:
129.11 g/mol
:
51 °C (1013 hPa)
:
1.199 g/cm³ (20 °C)
:
Ambient
:
226-853-0
:
1992
:
3,III
:
GHS02
:
GHS07