2,5-Difluoroaniline
Glass bottle for solids, 25 g
Catalog # MOLEM30567445
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:367-30-6
- Formula:C₆H₅F₂N
- MDL Number:MFCD00007651
- Molecular Weight:129.11 g/mol
- Boiling Pt:176 °C (1013 hPa)
- Melting Pt:13 °C
- Density:1.285 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:206-690-1
- UN:2941
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



