Order Entry
Export
ContactUsLinkComponent
2,4-Difluoroaniline
Irritant
2,4-Difluoroaniline
Catalog # MOLEM39956833
Supplier:  Molekula
CAS Number:  
Irritant
2,4-Difluoroaniline
Catalog # MOLEM39956833
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :129.11 g/mol
Merck Index:13,03171
Boiling Pt:170 °C (1013 hPa)
Melting Pt:–7 °C
Density:1.268 g/cm³ (20 °C)
Flash Pt:62 °C
Storage Temperature:Ambient
EINECS:206-687-5
UN:2941
ADR:6.1,III
 :GHS07
CAS Number:367-25-9
Formula:C₆H₅F₂N
MDL Number:MFCD00007648