Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Difluoroaniline

Irritant
Supplier:   Thermo Scientific
2,4-Difluoroaniline
Supplier:  Thermo Scientific
Certificates
2,4-Difluoroaniline

3 Options Available Below

Product Details & Documents
  • CAS Number:367-25-9
  • Formula:C₆H₅F₂N
  • MDL Number:MFCD00007648
  • Molecular Weight:129.11 g/mol
  • Merck Index:13,03171
  • Boiling Pt:170 °C (1013 hPa)
  • Melting Pt:–7 °C
  • Density:1.268 g/cm³ (20 °C)
  • Flash Pt:62 °C
  • Storage Temperature:Ambient
  • EINECS:206-687-5
  • UN:2941
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g