2,3-Difluoroaniline
Catalog # MOLEM87457736
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:129.11 g/mol
Boiling Pt:69 °C (12 mmHg)
Density:1.274 g/cm³ (20 °C)
EINECS:224-847-2
UN:2941
ADR:6.1,III
CAS Number:4519-40-8
Formula:C₆H₅F₂N
MDL Number:MFCD00010298