
1,2-Difluoro-4-iodobenzene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:64248-58-4
:
Formula:C₆H₃F₂I
MDL Number:MFCD00044630
:239.99 g/mol
Boiling Pt:177 °C (10 mmHg)
Density:1,99 g/cm³ (20 °C)
:GHS07
Storage Temperature:Refrigerator