
3,4-Difluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:455-86-7
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD00011672
:158.1 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:121…124 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-249-6
:GHS07