
2,4-Difluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1583-58-0
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD00011670
:158.1 g/mol
Boiling Pt:241 °C (1013 hPa)
Melting Pt:186…190 °C
Storage Temperature:Ambient
EINECS:216-430-9
:GHS07