

3,5-Difluorobenzonitrile ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64248-63-1
:
Formula:C₇H₃F₂N
MDL Number:MFCD00010311
:139.1 g/mol
Boiling Pt:160 °C
Melting Pt:83…88 °C
Storage Temperature:Ambient
EINECS:264-752-3
UN:3439
ADR:6.1,III
:GHS06
:GHS07