Order Entry
Export
ContactUsLinkComponent
 
Toxic
Irritant
2,5-Difluorobenzonitrile ≥98%
Catalog # A14014.06
CAS Number:  
Toxic
Irritant
2,5-Difluorobenzonitrile ≥98%
Catalog # A14014.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :139.1 g/mol
Boiling Pt:188 °C
Melting Pt:31…35 °C
Flash Pt:77 °C (170 °F)
Storage Temperature:Ambient
EINECS:264-753-9
UN:3439
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:64248-64-2
Formula:C₇H₃F₂N
MDL Number:MFCD00001777