2,4-Difluorobenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 3939-09-1 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 110°(230°F) Odour: Almond-like Boiling Point: 189° Beilstein Reference: 1940316 UN No.: 3439 MDL No.: MFCD00009826
- CAS Number:3939-09-1
- Formula:C₇H₃F₂N
- MDL Number:MFCD00009826
- Molecular Weight:139.1 g/mol
- Boiling Pt:189 °C
- Melting Pt:43…48 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:223-523-8
- UN:3439
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 3939-09-1 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 110°(230°F) Odour: Almond-like Boiling Point: 189° Beilstein Reference: 1940316 UN No.: 3439 MDL No.: MFCD00009826
- CAS Number:3939-09-1
- Formula:C₇H₃F₂N
- MDL Number:MFCD00009826
- Molecular Weight:139.1 g/mol
- Boiling Pt:189 °C
- Melting Pt:43…48 °C
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:223-523-8
- UN:3439
- ADR:6.1,III



