Order Entry
Export
ContactUsLinkComponent
 
 
 
3,5-Difluorobenzaldehyde 98+%
 
undefined
 
 
3,5-Difluorobenzaldehyde 98+%

 

  • Appearance:
    Clear light green Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4930 to 1.4950 (20°C, 589 nm)
  • GC:
    ≥98.0 %