Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Difluorobenzaldehyde

Supplier:   Thermo Scientific
3,4-Difluorobenzaldehyde
Supplier:  Thermo Scientific
Certificates
3,4-Difluorobenzaldehyde

3 Options Available Below

Product Details & Documents
  • CAS Number:34036-07-2
  • Formula:C₇H₄F₂O
  • MDL Number:MFCD00010328
  • Molecular Weight:142.1 g/mol
  • Boiling Pt:53 °C (1013 hPa)
  • Density:1,29 g/cm³ (20 °C)
  •  :Density: 1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:422-180-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g