
3,4-Difluorobenzaldehyde 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34036-07-2
:
Formula:C₇H₄F₂O
MDL Number:MFCD00010328
:142.1 g/mol
Boiling Pt:53 °C (1013 hPa)
Density:1.29 g/cm³
Flash Pt:65 °C
EINECS:422-180-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4990 to 1.5010 (20°C, 589 nm)
- GC:≥95 %