2,6-Difluorobenzaldehyde
Glass bottle for solids, 5 g
Catalog # MOLE23482755-5G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:437-81-0
- Formula:C₇H₄F₂O
- MDL Number:MFCD00010293
- Molecular Weight:142.1 g/mol
- Boiling Pt:194 °C (1013 hPa)
- Melting Pt:15…17 °C
- Density:1,31 g/cm³ (20 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



