Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluorobenzaldehyde
2,6-Difluorobenzaldehyde ≥97%
2,6-Difluorobenzaldehyde ≥97%
:
Thermo Fisher Scientific
2,6-Difluorobenzaldehyde ≥97%
:
Thermo Fisher Scientific
:
437-81-0
:
:
C₇H₄F₂O
:
MFCD00010293
:
85…86 °C (20 mmHg)
:
15…17 °C
:
1.317
:
73 °C (163 °F)
:
142.1 g/mol
:
GHS07
:
Ambient
msds_ALFAA18099
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years