Order Entry
Export
ContactUsLinkComponent
 
 
 
2,5-Difluorobenzaldehyde 98%
 
undefined
 
 
2,5-Difluorobenzaldehyde 98%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4970 to 1.5010 (20°C, 589 nm)
  • GC:
    ≥97.5 %