

2,4-Difluorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1550-35-2
:
Formula:C₇H₄F₂O
MDL Number:MFCD00010326
:142.1 g/mol
Boiling Pt:65…66 °C (17 mmHg)
Melting Pt:2…3 °C
Density:1.299
Flash Pt:55 °C (131 °F)
Storage Temperature:Ambient
EINECS:216-287-2
UN:1989
ADR:3,III
:GHS02
:GHS07