Order Entry
Export
ContactUsLinkComponent
2,4-Difluorobenzaldehyde 98%
Irritant
Flammable
2,4-Difluorobenzaldehyde 98%
Certificates
undefined
Irritant
Flammable
2,4-Difluorobenzaldehyde 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4970 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information