Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluorobenzaldehyde
2,3-Difluorobenzaldehyde
2,3-Difluorobenzaldehyde
MOLE19184103-5G
:
Molekula
:
2,3-Difluorobenzaldehyde
MOLE19184103-5G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
142.1 g/mol
:
64…65 °C (17 mmHg)
:
1.3 g/cm³ (20 °C)
:
1989
:
3,III
:
GHS07
:
2646-91-5
:
C₇H₄F₂O
:
MFCD00010292