1,3-Difluorobenzene
Glass bottle for solids, 100 g
Catalog # MOLE13117700-100G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:372-18-9
- Formula:C₆H₄F₂
- MDL Number:MFCD00000327
- Molecular Weight:114.09 g/mol
- Boiling Pt:83 °C (1013 hPa)
- Melting Pt:–59 °C
- Density:1.16 g/cm³ (20 °C)
- Flash Pt:2 °C
- Storage Temperature:Ambient
- EINECS:206-746-5
- UN:1993
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



