Order Entry
Export
ContactUsLinkComponent
 
 
1,3-Difluorobenzene 99+%
 
undefined
 
1,3-Difluorobenzene 99+%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4370 to 1.4390 (20°C, 589 nm)
  • GC:
    ≥99.0 %