1,2-Difluorobenzene
Catalog # MOLEM71909834
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:114.09 g/mol
Boiling Pt:92 °C (1013 hPa)
Melting Pt:–34 °C
Density:1.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-680-7
UN:1993
ADR:3,II
CAS Number:367-11-3
Formula:C₆H₄F₂
MDL Number:MFCD00000284