
1,2-Difluorobenzene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:367-11-3
:
Formula:C₆H₄F₂
MDL Number:MFCD00000284
:114.09 g/mol
Boiling Pt:92 °C (1013 hPa)
Melting Pt:–34 °C
Density:1.158 g/cm³
Flash Pt:2 °C
Storage Temperature:Ambient
EINECS:206-680-7
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4417 to 1.4437 (20°C, 589 nm)
- GC:≥97.5 %