0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Dichlorothiophenol
2,4-Dichlorothiophenol ≥97%
2,4-Dichlorothiophenol ≥97%
:
Thermo Fisher Scientific
2,4-Dichlorothiophenol ≥97%
:
Thermo Fisher Scientific
:
1122-41-4
:
:
C₆H₄Cl₂S
:
MFCD00041421
:
179.07 g/mol
:
60…62 °C (1.5 mmHg)
:
∼15 °C
:
1.440
:
>110 °C (230 °F)
:
214-346-7
:
GHS07
:
Ambient
msds_ALFAA12384
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years