Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dichlorothiophenol
3,4-Dichlorothiophenol ≥97%
3,4-Dichlorothiophenol ≥97%
:
Thermo Fisher Scientific
3,4-Dichlorothiophenol ≥97%
:
Thermo Fisher Scientific
:
5858-17-3
:
:
C₆H₄Cl₂S
:
MFCD00004840
:
179.07 g/mol
:
119…120 °C (10 mmHg)
:
87…89 °C
:
1.407
:
>110 °C (230 °F)
:
227-485-3
:
GHS07
:
Ambient
msds_ALFAL12031
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years