Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dichlorothiophenol

Supplier:   Thermo Scientific
2,3-Dichlorothiophenol
Supplier:  Thermo Scientific
Certificates
2,3-Dichlorothiophenol

2 Options Available Below

Product Details & Documents
Synonyms:  2,3-Dichlorobenzenethiol, 1,2-Dichloro-3-mercaptobenzene, 2,3-Dichlorobenzene-1-thiol
  • CAS Number:17231-95-7
  • Formula:C₆H₄Cl₂S
  • MDL Number:MFCD00041420
  • Molecular Weight:179.07 g/mol
  • Boiling Pt:116…118 °C (20 mmHg)
  • Melting Pt:46,5 °C
  • Melting Pt:46.5 °C
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g