2,4,6-Trichlorophenol 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:88-06-2
- Formula:C₆H₃Cl₃O
- MDL Number:MFCD00002172
- Molecular Weight:197.45 g/mol
- Merck Index:12,09773
- Boiling Pt:246 °C (760 mmHg)
- Melting Pt:64…68 °C
- Density:1,675 g/cm³ (25 °C)
- Flash Pt:99 °C
- Storage Temperature:Ambient
- EINECS:201-795-9
- UN:2020
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:88-06-2
- Formula:C₆H₃Cl₃O
- MDL Number:MFCD00002172
- Molecular Weight:197.45 g/mol
- Merck Index:12,09773
- Boiling Pt:246 °C (760 mmHg)
- Melting Pt:64…68 °C
- Density:1,675 g/cm³ (25 °C)
- Flash Pt:99 °C
- Storage Temperature:Ambient
- EINECS:201-795-9
- UN:2020
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:Light beige to light brown Crystalline powder or mass and/or chunks
- Infrared spectrum:Conforms
- Melting point:64 to 68 °C
- GC:≥97.5 %



