Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4,5-Tetrachlorophenol
2,3,4,5-Tetrachlorophenol
2,3,4,5-Tetrachlorophenol
EHERC17374500
:
EHRENSTORFER
:
2,3,4,5-Tetrachlorophenol
EHERC17374500
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
231.89 g/mol
:
289 °C (1013 hPa)
:
116 °C
:
1.6 g/cm³ (20 °C)
:
225-531-7
:
2020
:
6.1,III
:
4901-51-3
:
C₆H₂Cl₄O