Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichlorophenol
2,5-Dichlorophenol
2,5-Dichlorophenol
EHERC12452000
:
EHRENSTORFER
:
2,5-Dichlorophenol
EHERC12452000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
163 g/mol
:
211 °C (1013 hPa)
:
60…61 °C
:
Ambient
:
209-520-4
:
2020
:
6.1,III
:
GHS07
:
GHS09
:
583-78-8
:
C₆H₄Cl₂O
:
MFCD00002174