
2,6-Dichlorophenol
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:87-65-0
:
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002176
:163 g/mol
Merck Index:13,03099
Boiling Pt:∼219 °C (1013 hPa)
Melting Pt:68 °C
Density:1.653 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-761-3
UN:2020
ADR:6.1,III
:GHS07