2,4-Dichloroanisole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:553-82-2
:
Formula:C₇H₆Cl₂O
MDL Number:MFCD00044772
:177.03 g/mol
Boiling Pt:125 °C (10 mmHg)
Melting Pt:24…27 °C
Density:1.288
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:209-051-5