Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-3,4-dinitrobenzene
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
:
Thermo Fisher Scientific
1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
:
Thermo Fisher Scientific
:
610-40-2
:
:
C₆H₃ClN₂O₄
:
MFCD00007212
:
202.55 g/mol
:
14…16 °C (4 mmHg)
:
∼20…25 °C
:
1.687
:
>110 °C (230 °F)
:
Ambient
:
210-223-7
:
1577
:
6.1,II
:
GHS06
:
GHS08
msds_ALFAB22773
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years