Order Entry
Export
ContactUsLinkComponent
1-Chloro-2,4-dinitrobenzene
Toxic
Health
Environment
1-Chloro-2,4-dinitrobenzene
Catalog # EHERC11405500
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
1-Chloro-2,4-dinitrobenzene
Catalog # EHERC11405500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :202.55 g/mol
Merck Index:13,02155
Boiling Pt:315 °C (1013 hPa)
Melting Pt:52…54 °C
Density:1.7 g/cm³ (20 °C)
Flash Pt:194 °C
EINECS:202-551-4
UN:3441
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:97-00-7
Formula:C₆H₃ClN₂O₄
MDL Number:MFCD00007075