Order Entry
Export
ContactUsLinkComponent
1-Chloro-2,4-dinitrobenzene 99%
Corrosive
Toxic
Environment
1-Chloro-2,4-dinitrobenzene 99%
Certificates
undefined
Corrosive
Toxic
Environment
1-Chloro-2,4-dinitrobenzene 99%

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder or crystals or mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 53 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information