Order Entry
Export
Orders LinkContactUsLinkComponent
4-Chloro-2-nitrophenol, Sigma-Aldrich®

4-Chloro-2-nitrophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIAL25580-250G
Supplier:   Merck
4-Chloro-2-nitrophenol, Sigma-Aldrich®
Catalog # SIAL25580-250G
Supplier:  Merck
CAS Number:  89-64-5
4-Chloro-2-nitrophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  89-64-5
Price
149,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:89-64-5
  • Formula:C₆H₄ClNO₃
  • MDL Number:MFCD00007113
  • Molecular Weight:173.56 g/mol
  • Melting Pt:85…88 °C
  • Storage Temperature:Ambient
  • EINECS:201-927-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications